Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DROSHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100412.1 | NP_001093882.1 | 5 | 1114 | ||
NM_013235.4 | NP_037367.3 | 4 | 1038 | |||
XM_005248291.3 | XP_005248348.1 | 5 | 1114 | |||
XM_005248292.3 | XP_005248349.1 | 5 | 1114 | |||
XM_005248293.3 | XP_005248350.1 | 5 | 1114 | |||
XM_005248294.3 | XP_005248351.1 | 5 | 1114 | |||
XM_011514033.2 | XP_011512335.1 | 5 | 1114 | |||
XM_017009399.1 | XP_016864888.1 | 5 | 1114 | |||
XM_017009400.1 | XP_016864889.1 | 5 | 1114 | |||
XM_017009401.1 | XP_016864890.1 | 5 | 1114 | |||
GenBank mRNA | AF189011.1 | - | 4 | 924 | ||
AJ242976.1 | - | 1 | 186 | |||
AK308093.1 | - | 4 | 923 | |||
BC054003.1 | - | 4 | 926 | 6148714 | ||
BX647724.1 | - | 5 | 1017 |