Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145290.3 | NP_660333.2 | 9 | 1383 | ||
XM_005248137.4 | XP_005248194.1 | 9 | 1387 | |||
XM_011513811.2 | XP_011512113.1 | 4 | 567 | |||
XM_017007834.1 | XP_016863323.1 | 9 | 909 | |||
GenBank mRNA | AK095866.1 | - | 4 | 565 | ||
AK299168.1 | - | 6 | 670 | |||
AY181243.1 | - | 7 | 738 | |||
AY358306.1 | - | 9 | 1326 | |||
BC026009.1 | - | 4 | 467 | |||
BC035645.1 | - | 9 | 1394 |
Set Membership: |
G-protein coupled receptor |