Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145290.3 | NP_660333.2 | 11 | 1866 | ||
XM_005248137.4 | XP_005248194.1 | 11 | 1870 | |||
XM_011513811.2 | XP_011512113.1 | 6 | 1050 | |||
XM_017007834.1 | XP_016863323.1 | 11 | 1392 | |||
GenBank mRNA | AK095866.1 | - | 6 | 1048 | ||
AK299168.1 | - | 8 | 1153 | |||
AL832257.1 | - | 3 | 335 | |||
AY181243.1 | - | 9 | 1221 | |||
AY358306.1 | - | 11 | 1809 | |||
BC026009.1 | - | 6 | 950 | |||
BC035645.1 | - | 11 | 1877 |
Set Membership: |
G-protein coupled receptor |