Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261890.4 | XP_005261947.1 | 8 | 1393 | ||
XM_006724392.3 | XP_006724455.1 | 12 | 2101 | |||
XM_006724395.3 | XP_006724458.1 | 9 | 1553 | |||
XM_006724396.2 | XP_006724459.1 | 5 | 931 | |||
XM_017029179.1 | XP_016884668.1 | 13 | 2782 | |||
XM_017029180.1 | XP_016884669.1 | 12 | 2240 | |||
XM_017029181.1 | XP_016884670.1 | 12 | 2678 | |||
XM_017029183.1 | XP_016884672.1 | 8 | 1369 | |||
XM_017029184.1 | XP_016884673.1 | 5 | 941 | |||
XM_017029185.1 | XP_016884674.1 | 15 | 3563 | |||
XM_017029186.1 | XP_016884675.1 | 18 | 3164 | |||
GenBank mRNA | M30474.1 | - | 5 | 646 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 13 | 1952 | |||
GenBank mRNA | BC143607.1 | - | 9 | 1698 |
Gene Symbol: LOC102724197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724921.3 | XP_006724984.1 | 12 | 2544 | ||
XM_006724924.3 | XP_006724987.1 | 11 | 2386 | |||
XM_006724928.3 | XP_006724991.1 | 7 | 1582 | |||
XM_006724929.2 | XP_006724992.1 | 5 | 931 | |||
XM_011546256.2 | XP_011544558.1 | 14 | 2597 | |||
XM_017030133.1 | XP_016885622.1 | 13 | 2109 | |||
XM_017030134.1 | XP_016885623.1 | 13 | 2112 | |||
XM_017030135.1 | XP_016885624.1 | 13 | 2537 | |||
XM_017030136.1 | XP_016885625.1 | 14 | 2579 | |||
XM_017030137.1 | XP_016885626.1 | 14 | 2576 | |||
XM_017030138.1 | XP_016885627.1 | 14 | 2099 | |||
XM_017030139.1 | XP_016885628.1 | 14 | 2102 | |||
XM_017030140.1 | XP_016885629.1 | 15 | 2569 | |||
XM_017030141.1 | XP_016885630.1 | 15 | 2566 | |||
XM_017030142.1 | XP_016885631.1 | 13 | 2410 | |||
XM_017030143.1 | XP_016885632.1 | 13 | 2464 | |||
XM_017030144.1 | XP_016885633.1 | 8 | 1473 | |||
XM_017030145.1 | XP_016885634.1 | 9 | 2106 |