Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166212.1 | NP_001159684.1 | 3 | 1046 | ||
NM_013246.2 | NP_037378.1 | 3 | 1127 | |||
GenBank mRNA | AF172854.1 | - | 3 | 976 | ||
BC012939.1 | - | 3 | 1003 | 4453813 |