Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282757.1 | NP_001269686.1 | 3 | 648 | ||
NM_001282758.1 | NP_001269687.1 | 3 | 437 | |||
NM_001282759.1 | NP_001269688.1 | 4 | 738 | |||
NM_013293.4 | NP_037425.1 | 2 | 347 | |||
XM_005249725.1 | XP_005249782.1 | 3 | 634 | |||
XM_011515331.1 | XP_011513633.1 | 2 | 338 | |||
XM_017012089.1 | XP_016867578.1 | 2 | 879 | |||
XM_017012090.1 | XP_016867579.1 | 4 | 689 | |||
GenBank mRNA | AB097048.1 | - | 2 | 288 | ||
AK298815.1 | - | 4 | 699 | |||
AK300565.1 | - | 3 | 590 | |||
AK300741.1 | - | 3 | 377 | |||
AK310719.1 | - | 2 | 879 | |||
AK311687.1 | - | 3 | 599 | |||
AK314801.1 | - | 2 | 288 | |||
BC017094.2 | - | 2 | 258 | 3907327 | ||
BG722500.1 | - | 2 | 351 | |||
BP376263.1 | - | 2 | 338 | |||
DB445598.1 | - | 2 | 345 | |||
U53209.1 | - | 2 | 287 |