Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307986.1 | NP_001294915.1 | 3 | 968 | ||
NM_013305.5 | NP_037437.2 | 2 | 860 | |||
XM_017025715.1 | XP_016881204.1 | 4 | 356 | |||
XM_017025716.1 | XP_016881205.1 | 2 | 501 | |||
GenBank mRNA | AK056270.1 | - | 2 | 860 | ||
AK130126.1 | - | 4 | 415 | |||
AK293608.1 | - | 3 | 608 | |||
AK310764.1 | - | 3 | 609 | |||
BC108910.1 | - | 2 | 410 | 40011279 | ||
BC108911.1 | - | 2 | 410 | 40011281 | ||
CR457037.1 | - | 2 | 199 | |||
U91641.1 | - | 2 | 246 |