Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006240.2 | NP_006231.2 | 5 | 574 | ||
NM_152224.1 | NP_689410.1 | 5 | 574 | |||
NM_152226.1 | NP_689412.1 | 5 | 574 | |||
XM_005274553.2 | XP_005274610.1 | 7 | 901 | |||
GenBank mRNA | AF023455.1 | - | 2 | 175 | ||
AF027977.1 | - | 2 | 327 | |||
AK290463.1 | - | 2 | 229 | |||
AK292235.1 | - | 5 | 573 | |||
AK295504.1 | - | 1 | 64 | |||
AK315809.1 | - | 2 | 190 | |||
BC036026.1 | - | 5 | 573 | 5273322 | ||
X97867.1 | - | 5 | 576 |