Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100176.1 | NP_001093646.1 | 5, 6 | 566 | ||
NM_013312.2 | NP_037444.2 | 5, 6 | 566 | |||
XM_005259873.2 | XP_005259930.1 | 5, 6 | 566 | |||
XM_005259874.2 | XP_005259931.1 | 5, 6 | 566 | |||
XM_005259876.4 | XP_005259933.1 | 5, 6 | 562 | |||
XM_005259877.4 | XP_005259934.1 | 5, 6 | 512 | |||
XM_011527943.2 | XP_011526245.1 | 5, 6 | 1548 | |||
XM_011527944.2 | XP_011526246.1 | 5, 6 | 1547 | |||
XM_011527945.2 | XP_011526247.1 | 5, 6 | 1514 | |||
XM_011527946.2 | XP_011526248.1 | 5, 6 | 1539 | |||
XM_017026720.1 | XP_016882209.1 | 5, 6 | 536 | |||
XM_017026721.1 | XP_016882210.1 | 5, 6 | 562 | |||
GenBank mRNA | AF044924.1 | - | 5, 6 | 469 | ||
BC012443.1 | - | 5, 6 | 489 | 3859634 |