Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012464.4 | NP_036596.3 | 21 | 4704 | ||
XM_011532214.1 | XP_011530516.1 | 21 | 4762 | |||
XM_017008570.1 | XP_016864059.1 | 22 | 4830 | |||
GenBank mRNA | AF282732.1 | - | 21 | 4308 | ||
BC136429.1 | - | 21 | 4614 | 9020418 | ||
BC136430.1 | - | 21 | 4614 | 9020419 | ||
BC144085.1 | - | 22 | 4683 | 9052605 |