Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012473.3 | NP_036605.2 | 3 | 430 | ||
XM_005261508.1 | XP_005261565.1 | 3 | 815 | |||
XM_006724226.1 | XP_006724289.1 | 3 | 551 | |||
GenBank mRNA | AF276920.1 | - | 2 | 353 | ||
AF480262.1 | - | 3 | 430 | |||
AK130187.1 | - | 1108 | ||||
AK301837.1 | - | 3 | 821 | |||
BC013726.1 | - | 3 | 391 | 2900137 | ||
BC050610.1 | - | 3 | 404 | 6093567 | ||
CR456601.2 | - | 3 | 468 | |||
CR541896.1 | - | 2 | 353 | |||
CR541917.1 | - | 2 | 353 | |||
U78678.1 | - | 2 | 353 |