Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012479.3 | NP_036611.2 | 2 | 2085 | ||
GenBank mRNA | AB024334.1 | - | 2 | 2057 | ||
AK024230.1 | - | 2 | 2070 | |||
BC020963.2 | - | 2 | 2082 | 3915246 |