Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013248.2 | NP_037380.1 | 2 | 953 | ||
XM_011529230.2 | XP_011527532.1 | 2 | 722 | |||
GenBank mRNA | AF156957.1 | - | 2 | 837 | ||
AK026360.1 | - | 2 | 931 | |||
BC000759.2 | - | 2 | 841 | 2820775 | ||
BC002687.2 | - | 2 | 808 | 3608037 | ||
BC003029.1 | - | 2 | 841 | 2820775 | ||
BC003410.1 | - | 2 | 953 | 3450767 |