Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127384.2 | NP_001120856.1 | 15 | 2159 | ||
NM_013266.3 | NP_037398.2 | 15 | 2206 | |||
XM_017016151.1 | XP_016871640.1 | 16 | 2201 | |||
XM_017016152.1 | XP_016871641.1 | 18 | 2498 | |||
XM_017016153.1 | XP_016871642.1 | 16 | 2209 | |||
XM_017016154.1 | XP_016871643.1 | 11 | 1577 | |||
XM_017016155.1 | XP_016871644.1 | 11 | 1587 | |||
XM_017016156.1 | XP_016871645.1 | 11 | 1464 | |||
XM_017016157.1 | XP_016871646.1 | 9 | 1614 | |||
GenBank mRNA | AF091606.1 | - | 15 | 2184 | ||
AK289756.1 | - | 15 | 2095 | |||
AK313882.1 | - | 15 | 2209 |