Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013281.3 | NP_037413.1 | 2 | 674 | ||
NM_198391.2 | NP_938205.1 | 3 | 868 | |||
XM_005260682.4 | XP_005260739.1 | 3 | 1386 | |||
XM_011529204.2 | XP_011527506.1 | 4 | 1523 | |||
XM_011529205.2 | XP_011527507.1 | 4 | 1185 | |||
GenBank mRNA | AB040902.1 | - | 2 | 562 | ||
AF169677.1 | - | 2 | 571 | |||
AK027297.1 | - | 2 | 585 | |||
AK027694.1 | - | 1 | 303 | |||
AK074883.1 | - | 2 | 615 | |||
AK074909.1 | - | 2 | 543 | |||
AL529481.3 | - | 3 | 824 | |||
AY358319.1 | - | 1 | 408 | |||
BC020870.1 | - | 2 | 581 | 4606852 | ||
CD654700.1 | - | 2 | 635 |