Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316676.1 | NP_001303605.1 | 6 | 673 | ||
NM_001316677.1 | NP_001303606.1 | 4 | 346 | |||
NM_001323354.1 | NP_001310283.1 | 4 | 2066 | |||
NM_001323355.1 | NP_001310284.1 | 4 | 479 | |||
NM_001323356.1 | NP_001310285.1 | 3, 4 | 473 | |||
NM_001323357.1 | NP_001310286.1 | 2 | 149 | |||
NM_006504.5 | NP_006495.1 | 5 | 502 | |||
NM_130435.4 | NP_569119.1 | 2 | 149 | |||
XM_005252691.2 | XP_005252748.1 | 4 | 346 | |||
XM_011539994.2 | XP_011538296.1 | 4 | 464 | |||
XM_011539995.1 | XP_011538297.1 | 4 | 464 | |||
XM_011539996.1 | XP_011538298.1 | 4 | 464 | |||
XM_011539998.1 | XP_011538300.1 | 4 | 464 | |||
XM_017016467.1 | XP_016871956.1 | 4 | 464 | |||
XM_017016468.1 | XP_016871957.1 | 4 | 253 | |||
XM_017016469.1 | XP_016871958.1 | 4 | 464 | |||
GenBank mRNA | AF406557.1 | - | 5 | 437 | ||
AJ315969.1 | - | 2 | 58 | |||
AJ430580.1 | - | 2 | 67 | |||
AK056111.1 | - | 4 | 2066 | |||
AK290414.1 | - | 2 | 85 | |||
AK291828.1 | - | 4 | 346 | |||
AK299498.1 | - | 4 | 315 | |||
BC050062.1 | - | 6 | 581 | 5274802 | ||
BE246451.1 | - | 2 | 130 | |||
BI559814.1 | - | 5 | 509 | |||
BI910569.1 | - | 2 | 112 | |||
BU159075.1 | - | 2 | 111 | |||
BX648180.1 | - | 2 | 155 | |||
DA470230.1 | - | 2 | 149 | |||
U36623.1 | - | 85 | ||||
X54134.1 | - | 5 | 274 |