Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM216A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013300.2 | NP_037432.2 | 5, 6 | 1076 | ||
XM_005253875.4 | XP_005253932.1 | 5, 6 | 586 | |||
XM_011538246.2 | XP_011536548.1 | 5, 6 | 604 | |||
GenBank mRNA | AK225731.1 | - | 5, 6 | 1067 | ||
BC020967.1 | - | 5, 6 | 892 | 3848037 | ||
CR457001.1 | - | 5, 6 | 627 | |||
U79274.1 | - | 5, 6 | 1043 |