Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100176.1 | NP_001093646.1 | 22 | 2566 | ||
NM_013312.2 | NP_037444.2 | 23 | 2572 | |||
XM_005259873.2 | XP_005259930.1 | 23 | 2569 | |||
XM_005259874.2 | XP_005259931.1 | 22 | 2563 | |||
XM_005259876.4 | XP_005259933.1 | 23 | 2568 | |||
XM_005259877.4 | XP_005259934.1 | 23 | 2518 | |||
XM_011527943.2 | XP_011526245.1 | 23 | 3554 | |||
XM_011527944.2 | XP_011526246.1 | 23 | 3553 | |||
XM_011527945.2 | XP_011526247.1 | 23 | 3520 | |||
XM_011527946.2 | XP_011526248.1 | 23 | 3545 | |||
XM_011527947.2 | XP_011526249.1 | 19 | 2123 | |||
XM_017026720.1 | XP_016882209.1 | 23 | 2542 | |||
XM_017026721.1 | XP_016882210.1 | 22 | 2562 | |||
GenBank mRNA | AF044924.1 | - | 23 | 2475 | ||
BC012443.1 | - | 22 | 2489 | 3859634 |