Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013319.2 | NP_037451.1 | 1, 2 | 868 | ||
XM_006710590.3 | XP_006710653.1 | 1, 2 | 868 | |||
GenBank mRNA | AF117064.1 | - | 1, 2 | 866 | ||
AK074890.1 | - | 1, 2 | 860 | |||
AK222808.1 | - | 1, 2 | 868 | |||
BC004468.2 | - | 1, 2 | 560 | 3936638 | ||
BT006832.1 | - | 1, 2 | 534 |