Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199835.1 | NP_001186764.1 | 7 | 1797 | ||
NM_001199837.1 | NP_001186766.1 | 5 | 1602 | |||
NM_001199838.1 | NP_001186767.1 | 4 | 1501 | |||
NM_001318198.1 | NP_001305127.1 | 8 | 1871 | |||
NM_001318199.1 | NP_001305128.1 | 7 | 1672 | |||
NM_013322.2 | NP_037454.2 | 7 | 1750 | |||
XM_006715712.2 | XP_006715775.1 | 7 | 1950 | |||
XM_017012085.1 | XP_016867574.1 | 9 | 1965 | |||
XM_017012086.1 | XP_016867575.1 | 9 | 2404 | |||
XM_017012087.1 | XP_016867576.1 | 8 | 1894 | |||
XM_017012088.1 | XP_016867577.1 | 8 | 1841 | |||
GenBank mRNA | AF121860.1 | - | 7 | 1663 | ||
BC031050.1 | - | 7 | 1644 | 5278015 | ||
BC034992.1 | - | 7 | 1723 | 4827599 | ||
BX648363.1 | - | 1 | 3210 |