Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF078859.1 | - | 3 | 262 | ||
AF078868.1 | - | 3 | 243 | |||
AF134478.1 | - | 3 | 218 | |||
AK027523.1 | - | 3 | 295 | |||
AK074710.1 | - | 2 | 193 | |||
AK225112.1 | - | 3 | 231 | |||
AK225138.1 | - | 3 | 271 | |||
AK296339.1 | - | 3 | 211 | |||
AL136546.1 | - | 2 | 150 | |||
AL833441.1 | - | 3 | 259 | |||
BC012842.1 | - | 3 | 332 | 3861915 | ||
BC013925.1 | - | 3 | 174 | 3685016 | ||
BC029376.1 | - | 3 | 211 | 4658276 | ||
BC091522.1 | - | 3 | 186 | 30513213 | ||
DQ250006.1 | - | 3 | 187 | |||
EB387625.1 | - | 3 | 393 |