Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NENF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013349.4 | NP_037481.1 | 1, 2 | 242 | ||
GenBank mRNA | AB126219.1 | - | 1, 2 | 185 | ||
AF049672.1 | - | 1, 2 | 40 | |||
AF173937.1 | - | 1, 2 | 245 | |||
AK074431.1 | - | 1, 2 | 210 | |||
AK223135.1 | - | 1, 2 | 210 | |||
AY762102.1 | - | 1, 2 | 201 | |||
BC008823.2 | - | 1, 2 | 191 | 4303614 |