Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308295.1 | - | 7 | 759 |
Gene Symbol: ZNF223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013361.5 | NP_037493.3 | 5 | 599 | ||
XM_017027258.1 | XP_016882747.1 | 5 | 844 | |||
XM_017027259.1 | XP_016882748.1 | 5 | 899 | |||
XM_017027260.1 | XP_016882749.1 | 2 | 1818 | |||
GenBank mRNA | AF187989.1 | - | 5 | 584 | ||
AK292327.1 | - | 5 | 562 | |||
BC022466.1 | - | 5 | 599 | 4793666 | ||
DR761157.1 | - | 3 | 426 |