Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013366.3 | NP_037498.1 | 2, 3 | 816 | ||
GenBank mRNA | AF191337.1 | - | 2, 3 | 743 | ||
AK294601.1 | - | 2, 3 | 771 | |||
BC001579.1 | - | 17 | ||||
BC032503.1 | - | 2, 3 | 748 | 5243107 |