Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013438.4 | NP_038466.2 | 1 | 717 | ||
NM_053067.2 | NP_444295.1 | 1 | 717 | |||
XM_005251948.2 | XP_005252005.1 | 1 | 447 | |||
GenBank mRNA | AF176069.3 | - | 1 | 143 | ||
AF293384.1 | - | 1 | 143 | |||
AK027556.1 | - | 1 | 416 | |||
AK027576.1 | - | 1 | 407 | |||
AK074535.1 | - | 1 | 669 | |||
AL136643.1 | - | 1 | 421 | |||
BC010066.2 | - | 1 | 397 | 4110398 | ||
BC039294.1 | - | 1 | 408 | 5263680 |