Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303007.1 | NP_001289936.1 | 1 | 1260 | ||
NM_001303008.1 | NP_001289937.1 | 2 | 360 | |||
NM_013974.2 | NP_039268.1 | 2 | 435 | |||
XM_011514448.2 | XP_011512750.1 | 2 | 511 | |||
GenBank mRNA | AF070667.1 | - | 2 | 432 | ||
AF087894.1 | - | 2 | 432 | |||
AK026191.1 | - | 1 | 787 | |||
AK098671.1 | - | 2 | 422 | |||
BC001435.2 | - | 2 | 347 | 3138832 | ||
BF338733.1 | - | 2 | 360 | |||
EB387244.1 | - | 2 | 334 | |||
FJ224290.1 | - | 2 | 432 |