Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002311.4 | NP_002302.2 | 2 | 464 | ||
NM_013975.3 | NP_039269.2 | 2 | 464 | |||
XM_005257970.3 | XP_005258027.1 | 2 | 432 | |||
XM_006721896.3 | XP_006721959.1 | 2 | 433 | |||
XM_017024624.1 | XP_016880113.1 | 2 | 628 | |||
GenBank mRNA | AK300363.1 | - | 2 | 432 | ||
AK308407.1 | - | 2 | 409 | |||
BC009026.1 | - | 2 | 385 | |||
BC068005.1 | - | 2 | 439 | 6092747 | ||
DB100562.1 | - | 2 | 464 | |||
U40671.1 | - | 2 | 425 | |||
X84740.1 | - | 2 | 407 |