Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014008.4 | NP_054727.1 | 3, 4 | 539 | ||
XM_005272599.3 | XP_005272656.1 | 3, 4 | 471 | |||
GenBank mRNA | AJ005890.1 | - | 3, 4 | 455 | ||
AK291976.1 | - | 3, 4 | 500 | |||
AK296911.1 | - | 3, 4 | 510 | |||
AK298264.1 | - | 3, 4 | 534 | |||
AK316375.1 | - | 3, 4 | 485 | |||
BC000972.2 | - | 3, 4 | 486 | 3449051 | ||
BC011675.2 | - | 3, 4 | 370 | 4299954 | ||
BG912845.1 | - | 3, 4 | 525 | |||
DC340647.1 | - | 3, 4 | 536 |