Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014033.3 | NP_054752.3 | 1 | 421 | ||
XM_006719332.1 | XP_006719395.1 | 2 | 775 | |||
GenBank mRNA | AF113007.1 | - | 1 | 95 | ||
AJ536233.1 | - | 1 | 143 | |||
AK023693.1 | - | 1 | 153 | |||
AK024409.1 | - | 1 | 186 | |||
AK225595.1 | - | 1 | 158 | |||
AK225689.1 | - | 1 | 157 | |||
AK293356.1 | - | 1 | 154 | |||
AK307071.1 | - | 1 | 161 | |||
AL050159.1 | - | 1 | 131 | |||
AY358797.1 | - | 1 | 189 | |||
BC004492.2 | - | 1 | 138 | 3689692 | ||
BC008180.2 | - | 1 | 139 | 2900478 | ||
BM805242.1 | - | 1 | 443 |