Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172129.1 | NP_001165600.1 | 3, 4 | 474 | ||
NM_001172132.1 | NP_001165603.1 | 4, 5 | 540 | |||
NM_001172133.1 | NP_001165604.1 | 4, 5 | 538 | |||
NM_002110.3 | NP_002101.2 | 3, 4 | 474 | |||
GenBank mRNA | AK026432.1 | - | 3, 4 | 408 | ||
AK225819.1 | - | 3, 4 | 420 | |||
AK290928.1 | - | 4, 5 | 486 | |||
AK298726.1 | - | 4, 5 | 484 | |||
BC014435.1 | - | 3, 4 | 360 | 4855747 | ||
BC094847.1 | - | 3, 4 | 371 | 5923744 | ||
BC108930.1 | - | 3, 4 | 401 | 40025497 | ||
BC108931.1 | - | 3, 4 | 401 | 40025500 | ||
BC114463.1 | - | 3, 4 | 401 | 40025502 | ||
DA465978.1 | - | 3, 4 | 475 | |||
M16592.1 | - | 2, 3 | 249 |