Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013339.3 | NP_037471.2 | 7 | 776 | ||
GenBank mRNA | AF063604.2 | - | 7 | 731 | ||
AF102851.1 | - | 7 | 709 | |||
AK022700.1 | - | 7 | 714 | |||
AK295286.1 | - | 1 | 22 | |||
BC001253.1 | - | 7 | 708 | 3452687 |