Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF078859.1 | - | 3 | 333 | ||
AF078868.1 | - | 3 | 314 | |||
AF134478.1 | - | 3 | 289 | |||
AK027523.1 | - | 3 | 366 | |||
AK074710.1 | - | 2 | 264 | |||
AK225112.1 | - | 3 | 302 | |||
AK225138.1 | - | 3 | 342 | |||
AK296339.1 | - | 3 | 282 | |||
AL136546.1 | - | 2 | 221 | |||
AL833441.1 | - | 3 | 330 | |||
BC012842.1 | - | 3 | 403 | 3861915 | ||
BC013925.1 | - | 3 | 245 | 3685016 | ||
BC029376.1 | - | 3 | 282 | 4658276 | ||
BC091522.1 | - | 3 | 257 | 30513213 | ||
DQ250006.1 | - | 3 | 258 | |||
EB387625.1 | - | 3 | 464 |