Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182981.2 | NP_892026.1 | 5 | 654 | ||
GenBank mRNA | AF191740.1 | - | 3 | 292 | ||
AK290383.1 | - | 5 | 636 | |||
AY037158.1 | - | 5 | 619 | |||
AY258066.1 | - | 6 | 1105 | |||
AY258067.1 | - | 5 | 945 | |||
AY258068.1 | - | 5 | 635 | |||
BC020434.1 | - | 5 | 620 | |||
BC032476.1 | - | 5 | 619 | 5215262 | ||
BC093687.1 | - | 5 | 537 | 7939532 | ||
BC113417.1 | - | 5 | 537 | 8322469 |