Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF119226.1 | - | 3 | 517 | ||
BC006286.1 | - | 3 | 538 | 3958403 | ||
BT006633.1 | - | 3 | 506 |