Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164595.1 | NP_001158067.1 | 10 | 2916 | ||
NM_013377.3 | NP_037509.3 | 8 | 2340 | |||
GenBank mRNA | AK091096.1 | - | 8 | 2307 | ||
AK094690.1 | - | 1 | 1179 | |||
AK131554.1 | - | 10 | 2516 | |||
AK294431.1 | - | 8 | 2396 | |||
AK294535.1 | - | 10 | 2379 | |||
AL133067.1 | - | 1 | 260 | |||
BC040922.1 | - | 4 | 1562 | |||
BC093744.1 | - | 8 | 2189 | 7939589 | ||
BC093746.2 | - | 8 | 2189 | 7939591 | ||
BX640695.1 | - | 6 | 2069 |