Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083335.1 | NP_001076804.1 | 4, 5 | 359 | ||
NM_013380.3 | NP_037512.3 | 3, 4 | 310 | |||
GenBank mRNA | AF198358.1 | - | 4, 5 | 302 | ||
AK023099.1 | - | 3, 4 | 280 | |||
AK302966.1 | - | 4, 5 | 417 | |||
BC101753.1 | - | 3, 4 | 254 | 8069259 | ||
BC117223.1 | - | 3, 4 | 254 | 40125774 | ||
DB288887.1 | - | 4, 5 | 359 |