Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039877.1 | NP_001034966.1 | 2 | 357 | ||
NM_013403.2 | NP_037535.2 | 2 | 357 | |||
XM_006723171.2 | XP_006723234.1 | 2 | 369 | |||
XM_006723172.2 | XP_006723235.1 | 2 | 369 | |||
XM_006723174.2 | XP_006723237.1 | 2 | 164 | |||
XM_011526878.1 | XP_011525180.1 | 2 | 369 | |||
XM_017026716.1 | XP_016882205.1 | 2 | 98 | |||
XM_017026717.1 | XP_016882206.1 | 2 | 119 | |||
XM_017026719.1 | XP_016882208.1 | 2 | 369 | |||
GenBank mRNA | AB209003.1 | - | 2 | 309 | ||
AF212940.1 | - | 2 | 324 | |||
AK297637.1 | - | 3 | 336 | |||
AK298804.1 | - | 3 | 264 | |||
BC034604.1 | - | 2 | 334 | 4829172 | ||
DA510367.1 | - | 2 | 357 |