Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013446.3 | NP_038474.2 | 7 | 1337 | ||
NR_117084.1 | 5 | 880 | ||||
XM_011515996.1 | XP_011514298.1 | 6 | 1126 | |||
XM_011515997.2 | XP_011514299.1 | 7 | 1394 | |||
XM_011515998.1 | XP_011514300.1 | 7 | 1079 | |||
GenBank mRNA | AB209298.1 | - | 6 | 1660 | ||
AF192784.1 | - | 7 | 1232 | |||
AK024987.1 | - | 2 | 82 | |||
AK027137.1 | - | 6 | 849 | |||
AK127030.1 | - | 8 | 1954 | |||
AK297361.1 | - | 5 | 679 | |||
AK315552.1 | - | 7 | 1136 | |||
AL136812.1 | - | 7 | 1235 | |||
BC025955.1 | - | 7 | 1270 | 4869484 | ||
BC037400.1 | - | 7 | 1122 | 5176176 | ||
BC064838.1 | - | 7 | 1115 | 6173715 |
Gene Symbol: MKRN9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033410.1 | 1 | 1257 |