Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014012.5 | NP_054731.2 | 5 | 1523 | ||
XM_005260404.1 | XP_005260461.1 | 5 | 1547 | |||
XM_011528795.1 | XP_011527097.1 | 5 | 1541 | |||
XM_017027833.1 | XP_016883322.1 | 5 | 1381 | |||
GenBank mRNA | AF152863.1 | - | 5 | 1505 | ||
BC039813.1 | - | 5 | 1509 | 5220518 |