Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM176B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101311.1 | NP_001094781.1 | 3 | 378 | ||
NM_001101312.1 | NP_001094782.1 | 2 | 370 | |||
NM_001101314.1 | NP_001094784.1 | 2 | 370 | |||
NM_014020.3 | NP_054739.3 | 2 | 502 | |||
XM_006715933.3 | XP_006715996.2 | 3 | 257 | |||
GenBank mRNA | AF115384.1 | - | 2 | 292 | ||
AK000841.1 | - | 2 | 201 | |||
AK097304.1 | - | 3 | 1339 | |||
AK129525.1 | - | 2 | 206 | |||
AK315575.1 | - | 3 | 188 | |||
BC004358.1 | - | 2 | 417 | 2822223 | ||
BC014586.2 | - | 2 | 417 | 2822223 | ||
BC091471.1 | - | 2 | 201 | |||
BI761752.1 | - | 2 | 370 | |||
BI910761.1 | - | 3 | 174 | |||
BP298052.1 | - | 2 | 502 | |||
DQ144974.1 | - | 2 | 260 | |||
R95048.1 | - | 2 | 298 |