Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166293.1 | NP_001159765.1 | 4 | 543 | ||
NM_001166294.1 | NP_001159766.1 | 3 | 411 | |||
NM_001166295.1 | NP_001159767.1 | 3 | 539 | |||
NM_001166417.1 | NP_001159889.1 | 5 | 721 | |||
NM_014021.3 | NP_054740.3 | 5 | 593 | |||
XM_005270427.1 | XP_005270484.1 | 4 | 482 | |||
XM_005270428.1 | XP_005270485.1 | 4 | 440 | |||
XM_005270429.1 | XP_005270486.1 | 4 | 482 | |||
XM_006710331.1 | XP_006710394.1 | 4 | 400 | |||
XM_011540610.1 | XP_011538912.1 | 5 | 490 | |||
XM_011540612.2 | XP_011538914.1 | 5 | 531 | |||
XM_017000234.1 | XP_016855723.1 | 4 | 471 | |||
XM_017000235.1 | XP_016855724.1 | 4 | 449 | |||
XM_017000236.1 | XP_016855725.1 | 4 | 465 | |||
XM_017000237.1 | XP_016855726.1 | 3 | 254 | |||
GenBank mRNA | AB023140.1 | - | 4 | 531 | ||
AK021515.1 | - | 2 | 1115 | |||
AK093173.1 | - | 5 | 478 | |||
AK292475.1 | - | 4 | 431 | |||
AK294057.1 | - | 3 | 298 | |||
AK316472.1 | - | 3 | 317 | |||
AL133046.1 | - | 5 | 533 | |||
AY367055.1 | - | 5 | 456 | |||
AY651262.1 | - | 2 | 116 | |||
BC033637.1 | - | 4 | 418 | 5578582 | ||
BC035580.1 | - | 3 | 352 | |||
BC064389.1 | - | 4 | 507 | 5745142 |