Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014026.4 | NP_054745.1 | 1 | 428 | ||
XM_011542778.2 | XP_011541080.1 | 1 | 136 | |||
XM_017017612.1 | XP_016873101.1 | 1 | 879 | |||
GenBank mRNA | AF077201.1 | - | 1 | 111 | ||
AF532613.1 | - | 1 | 428 | |||
AK026554.1 | - | 1 | 124 | |||
AK223089.1 | - | 1 | 124 | |||
AK307403.1 | - | 1 | 120 | |||
AY040771.1 | - | 1 | 126 | |||
AY077684.1 | - | 1 | 99 | |||
BC014532.1 | - | 1 | 121 | 4064215 | ||
BE382749.1 | - | 1 | 90 |