Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013337.2 | NP_037469.2 | 4 | 1603 | ||
GenBank mRNA | AF155330.1 | - | 4 | 1582 | ||
AK000830.1 | - | 4 | 1584 | |||
AK225453.1 | - | 5 | 1473 | |||
BC002324.2 | - | 4 | 1603 | 2819655 |