Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013366.3 | NP_037498.1 | 8 | 1617 | ||
GenBank mRNA | AB037827.1 | - | 6 | 793 | ||
AF191337.1 | - | 8 | 1544 | |||
AK294601.1 | - | 8 | 1572 | |||
BC001579.1 | - | 6 | 809 | |||
BC009487.2 | - | 4 | 408 | |||
BC032503.1 | - | 8 | 1549 | 5243107 |