Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013444.3 | NP_038472.2 | 1 | 3124 | ||
GenBank mRNA | AF189009.1 | - | 1 | 3082 | ||
AK001029.1 | - | 2 | 1691 | |||
AL442081.1 | - | 1 | 2737 | |||
AU310783.1 | - | 1 | 285 | |||
BC069237.1 | - | 2 | 2995 | 4543266 |