Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013451.3 | NP_038479.1 | 11 | 1062 | ||
NM_133337.2 | NP_579899.1 | 11 | 1062 | |||
XM_005269693.4 | XP_005269750.1 | 11 | 1241 | |||
XM_005269694.4 | XP_005269751.1 | 11 | 1241 | |||
XM_011539632.2 | XP_011537934.1 | 12 | 1435 | |||
XM_017016068.1 | XP_016871557.1 | 10 | 822 | |||
XM_017016069.1 | XP_016871558.1 | 10 | 867 | |||
XM_017016070.1 | XP_016871559.1 | 11 | 1249 | |||
GenBank mRNA | AB033033.2 | - | 11 | 939 | ||
AF182316.1 | - | 11 | 1035 | |||
AF207990.1 | - | 11 | 939 | |||
BC040110.1 | - | 12 | 1252 | |||
BC052617.1 | - | 11 | 1027 | 6422729 |