Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003373.3 | NP_003364.1 | 3, 4 | 492 | ||
NM_014000.2 | NP_054706.1 | 3, 4 | 492 | |||
GenBank mRNA | AK126978.1 | - | 3, 4 | 747 | ||
BC039174.1 | - | 3, 4 | 479 | 4520338 | ||
BX537994.1 | - | 5, 6 | 357 | |||
EU794673.1 | - | 3, 4 | 492 | |||
M33308.1 | - | 3, 4 | 448 |