Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014008.4 | NP_054727.1 | 15 | 1843 | ||
XM_005272599.3 | XP_005272656.1 | 15 | 1772 | |||
GenBank mRNA | AJ005890.1 | - | 15 | 1759 | ||
AK291976.1 | - | 15 | 1804 | |||
AK298264.1 | - | 15 | 1838 | |||
AK316375.1 | - | 15 | 1789 | |||
BC000972.2 | - | 15 | 1790 | 3449051 | ||
BC011675.2 | - | 15 | 1674 | 4299954 |