Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184734.1 | NP_001171663.1 | 9 | 1636 | ||
NM_014010.4 | NP_054729.3 | 22 | 4475 | |||
NM_198186.3 | NP_937829.3 | 8 | 2240 | |||
NM_198187.3 | NP_937830.3 | 9 | 1691 | |||
NM_198188.2 | NP_937831.1 | 9 | 1670 | |||
GenBank mRNA | AB014534.1 | - | 22 | 4475 | ||
AF116574.1 | - | 20 | 3696 | |||
AK024064.1 | - | 9 | 1641 | |||
AK127350.1 | - | 21 | 3696 | |||
BC029272.1 | - | 8 | 2240 | |||
BC146756.1 | - | 22 | 4475 | 9007192 | ||
BX649176.1 | - | 22 | 3810 |