Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014044.5 | NP_054763.2 | 2 | 270 | ||
XM_005263913.4 | XP_005263970.1 | 2 | 366 | |||
XM_006712403.3 | XP_006712466.1 | 2 | 366 | |||
XM_006712404.2 | XP_006712467.1 | 2 | 280 | |||
GenBank mRNA | AF077038.1 | - | 2 | 222 | ||
AK222599.1 | - | 2 | 236 | |||
AL080115.1 | - | 2 | 274 | |||
AY017215.1 | - | 2 | 238 | |||
AY251534.1 | - | 1 | 131 | |||
BC017019.1 | - | 2 | 403 | 3888349 | ||
BC025992.2 | - | 1 | 1298 | 4830706 | ||
BC039078.1 | - | 1 | 1274 | 5166015 | ||
BC091481.1 | - | 2 | 260 | 30337350 | ||
CR533464.1 | - | 1 | 118 | |||
KU178586.1 | - | 1 | 118 | |||
KU178587.1 | - | 1 | 118 |